siRNA Details
Gene Symbol: CALCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033953.2 | NP_001029125.1 | 5 | 593 | ||
NR_125898.1 | 6 | 741 | ||||
XM_017018283.1 | XP_016873772.1 | 5 | 1787 | |||
XM_017018284.1 | XP_016873773.1 | 5 | 1811 | |||
GenBank mRNA | BX094781.1 | - | 2 | 263 | ||
CB048118.1 | - | 2 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
