siRNA Details
Gene Symbol: ERICH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130514.1 | NP_001123986.1 | 2 | 493 | ||
XM_017026119.1 | XP_016881608.1 | 2 | 1288 | |||
GenBank mRNA | CB047448.1 | - | 1 | 82 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
