siRNA Details
Gene Symbol: SSTR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001053.3 | NP_001044.1 | 1 | 320 | ||
NM_001172560.1 | NP_001166031.1 | 2 | 303 | |||
GenBank mRNA | AK308381.1 | - | 1 | 320 | ||
DB202230.1 | - | 1 | 320 | |||
DQ448303.1 | - | 1 | 232 | |||
DQ448304.1 | - | 1 | 232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
