siRNA Details
Gene Symbol: ANKHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197030.1 | NP_001183959.1 | 2 | 529 | ||
NM_017747.2 | NP_060217.1 | 2 | 529 | |||
NM_017978.2 | NP_060448.1 | 2 | 529 | |||
NM_024668.3 | NP_078944.2 | 2 | 529 | |||
GenBank mRNA | AF258557.1 | - | 2 | 489 | ||
AF521882.1 | - | 2 | 435 | |||
AK022041.1 | - | 1 | 65 | |||
AK074173.1 | - | 2 | 493 | |||
AK098574.1 | - | 2 | 499 | |||
BC004457.1 | - | 2 | 474 | |||
BC009420.1 | - | 2 | 422 | |||
BC009909.2 | - | 2 | 474 | |||
BC117677.2 | - | 2 | 455 | |||
BC117678.2 | - | 2 | 455 | |||
BC127127.1 | - | 2 | 455 | |||
BC150486.2 | - | 2 | 459 | |||
DA509209.1 | - | 2 | 529 |
Gene Symbol: ANKHD1-EIF4EBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020690.5 | NP_065741.3 | 2 | 529 | ||
GenBank mRNA | AF521883.1 | - | 2 | 435 | ||
BC144623.1 | - | 2 | 469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
