siRNA Details
Gene Symbol: SSX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005636.3 | NP_005627.1 | 6 | 503 | ||
XM_017029759.1 | XP_016885248.1 | 6 | 561 | |||
GenBank mRNA | AX740218.1 | - | 5 | 445 | ||
BC137394.1 | - | 5 | 470 | 9021392 | ||
BC137395.1 | - | 5 | 470 | 9021393 | ||
U90841.1 | - | 5 | 445 |
Gene Symbol: SSX4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034832.3 | NP_001030004.1 | 6 | 514 | ||
XM_017029613.1 | XP_016885102.1 | 6 | 561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
