siRNA Details
Gene Symbol: HIST1H3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003536.2 | NP_003527.1 | 1 | 239 | ||
GenBank mRNA | AK311991.1 | - | 1 | 252 | ||
BC007518.2 | - | 1 | 367 | |||
BC096128.3 | - | 1 | 229 | 40002508 | ||
BC096129.3 | - | 1 | 229 | 40002509 | ||
BC096130.3 | - | 1 | 229 | 40002510 | ||
BC143046.1 | - | 1 | 229 | 40112088 | ||
BC148243.1 | - | 1 | 229 | 40082529 | ||
BC148250.1 | - | 1 | 229 | 40002516 |
Gene Symbol: HIST1H3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003530.4 | NP_003521.2 | 2 | 683 | ||
GenBank mRNA | AK316611.1 | - | 2 | 683 | ||
BC031333.1 | - | 2 | 602 | 3608479 | ||
BQ051491.1 | - | 2 | 626 |
Gene Symbol: HIST1H3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021018.2 | NP_066298.1 | 1 | 231 | ||
GenBank mRNA | AK314142.1 | - | 1 | 264 | ||
BC067492.1 | - | 1 | 229 | 7002073 | ||
BC069818.1 | - | 1 | 231 | 7262721 | ||
BC096131.2 | - | 1 | 229 | 40002512 | ||
CR541983.1 | - | 1 | 229 | |||
CR542011.1 | - | 1 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
