siRNA Details
Gene Symbol: WWOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291997.1 | NP_001278926.1 | 8 | 2257 | ||
NM_016373.3 | NP_057457.1 | 9 | 2322 | |||
XM_011523103.2 | XP_011521405.1 | 10 | 2157 | |||
XM_017023279.1 | XP_016878768.1 | 2 | 1787 | |||
GenBank mRNA | AF211943.1 | - | 9 | 2081 | ||
AF227527.1 | - | 9 | 2060 | |||
AF395123.1 | - | 6 | 1549 | |||
AF395124.1 | - | 5 | 1442 | |||
AK027626.1 | - | 9 | 2080 | |||
AY256821.1 | - | 9 | 2009 | |||
BC003184.1 | - | 6 | 1490 | 3535809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
