siRNA Details
Gene Symbol: TP53TG3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023955.1 | XP_016879444.1 | 1 | 1406 | ||
XM_017023956.1 | XP_016879445.1 | 1 | 1406 | |||
XM_017023959.1 | XP_016879448.1 | 1 | 1407 |
Gene Symbol: LOC102723655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724904.3 | XP_006724967.1 | 1 | 1411 | ||
XM_006724905.3 | XP_006724968.1 | 1 | 1407 | |||
XM_017030123.1 | XP_016885612.1 | 1 | 1406 | |||
XM_017030124.1 | XP_016885613.1 | 1 | 1406 |
Gene Symbol: TP53TG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023102.1 | XP_016878591.1 | 1 | 1406 |
Gene Symbol: TP53TG3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023638.1 | XP_016879127.1 | 1 | 1406 |
Gene Symbol: TP53TG3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023591.1 | XP_016879080.1 | 1 | 1406 |
Gene Symbol: TP53TG3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023633.1 | XP_016879122.1 | 1 | 309 | ||
XM_017023634.1 | XP_016879123.1 | 1 | 309 | |||
XM_017023635.1 | XP_016879124.1 | 1 | 310 | |||
XM_017023636.1 | XP_016879125.1 | 1 | 310 | |||
XM_017023637.1 | XP_016879126.1 | 1 | 310 |
Gene Symbol: TP53TG3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023950.1 | XP_016879439.1 | 1 | 1406 | ||
XM_017023951.1 | XP_016879440.1 | 1 | 1406 | |||
XM_017023954.1 | XP_016879443.1 | 1 | 1407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
