siRNA Details
Gene Symbol: AVP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000490.4 | NP_000481.2 | 1 | 48 | ||
XM_011529267.1 | XP_011527569.1 | 2 | 1578 | |||
GenBank mRNA | AF031475.1 | - | 1 | 736 | ||
BC126196.1 | - | 1 | 25 | 8991912 | ||
BC126224.1 | - | 1 | 25 | 8991940 | ||
BG715518.1 | - | 1 | 53 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
