siRNA Details
Gene Symbol: SULT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177552.3 | NP_808220.1 | 5 | 532 | ||
GenBank mRNA | AB111093.1 | - | 4 | 428 | ||
AB111094.1 | - | 4 | 428 | |||
AB111095.1 | - | 4 | 428 | |||
AB111096.1 | - | 4 | 428 | |||
AB111097.1 | - | 4 | 428 | |||
AB111098.1 | - | 4 | 428 | |||
AB111099.1 | - | 4 | 428 | |||
AK298073.1 | - | 5 | 533 | |||
BC078144.1 | - | 5 | 476 | 6022425 | ||
BC132914.1 | - | 5 | 529 | 40146936 | ||
BC132920.1 | - | 5 | 529 | 40146942 | ||
BC144354.1 | - | 5 | 529 | 9052874 | ||
BC144355.1 | - | 5 | 529 | 9052875 | ||
BC171772.1 | - | 5 | 529 | 9054426 | ||
DA978913.1 | - | 5 | 534 | |||
L25275.1 | - | 5 | 492 | |||
U08032.1 | - | 4 | 434 | |||
U34199.1 | - | 4 | 428 |
Gene Symbol: SLX1A-SULT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037608.1 | 8 | 1349 | |||
GenBank mRNA | AK094769.1 | - | 6 | 1831 | ||
AK097386.1 | - | 6 | 897 | |||
AK122707.1 | - | 8 | 1350 | |||
AK122733.1 | - | 9 | 1175 | |||
BC014471.1 | - | 6 | 614 | 4907290 | ||
BC111011.1 | - | 7 | 1377 | 5768190 | ||
X84653.1 | - | 5 | 449 |
Gene Symbol: SLX1B-SULT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037609.1 | 8 | 1349 |
Gene Symbol: SULT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017390.2 | NP_001017390.1 | 5 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
