siRNA Details
Gene Symbol: ZNF365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199452.3 | NP_955524.3 | 3 | 399 | ||
XM_017015938.1 | XP_016871427.1 | 2 | 405 | |||
XM_017015939.1 | XP_016871428.1 | 2 | 405 | |||
XM_017015940.1 | XP_016871429.1 | 2 | 372 | |||
XM_017015941.1 | XP_016871430.1 | 2 | 405 | |||
GenBank mRNA | AJ505150.1 | - | 3 | 399 | ||
BC044306.1 | - | 2 | 120 | |||
BC101082.1 | - | 2 | 218 | |||
BC101085.2 | - | 2 | 218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
