siRNA Details
Gene Symbol: GPATCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002909.3 | NP_001002909.1 | 8 | 3308 | ||
NM_001304939.1 | NP_001291868.1 | 7 | 3233 | |||
NM_001304940.1 | NP_001291869.1 | 10 | 3449 | |||
NM_001304941.1 | NP_001291870.1 | 9 | 3326 | |||
NM_001304942.1 | NP_001291871.1 | 7 | 3235 | |||
NM_001304943.1 | NP_001291872.1 | 5 | 3189 | |||
XM_011524557.1 | XP_011522859.1 | 10 | 3382 | |||
XM_011524558.1 | XP_011522860.1 | 11 | 3201 | |||
XM_011524559.2 | XP_011522861.1 | 12 | 3279 | |||
XM_011524561.2 | XP_011522863.1 | 8 | 3040 | |||
XM_017024372.1 | XP_016879861.1 | 13 | 3324 | |||
XM_017024373.1 | XP_016879862.1 | 10 | 4159 | |||
XM_017024374.1 | XP_016879863.1 | 12 | 3242 | |||
XM_017024375.1 | XP_016879864.1 | 9 | 3337 | |||
XM_017024376.1 | XP_016879865.1 | 10 | 3197 | |||
XM_017024377.1 | XP_016879866.1 | 9 | 3174 | |||
XM_017024378.1 | XP_016879867.1 | 10 | 3355 | |||
XM_017024379.1 | XP_016879868.1 | 11 | 3237 | |||
XM_017024380.1 | XP_016879869.1 | 10 | 3175 | |||
GenBank mRNA | AB011125.2 | - | 7 | 4398 | ||
BC024147.2 | - | 1 | 87 | |||
BC136629.1 | - | 9 | 3125 | 9020619 | ||
BC144323.1 | - | 9 | 3125 |
