siRNA Details
Gene Symbol: ZNF365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199450.2 | NP_955522.1 | 5 | 1677 | ||
GenBank mRNA | AJ505148.1 | - | 5 | 1639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
