siRNA Details
Gene Symbol: RPL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000985.4 | NP_000976.1 | 6 | 723 | ||
NM_001035006.2 | NP_001030178.1 | 6 | 474 | |||
NM_001199340.1 | NP_001186269.1 | 7 | 572 | |||
NM_001199341.1 | NP_001186270.1 | 6 | 573 | |||
NM_001199342.1 | NP_001186271.1 | 7 | 671 | |||
NM_001199343.1 | NP_001186272.1 | 7 | 654 | |||
NM_001199344.1 | NP_001186273.1 | 6 | 556 | |||
NM_001199345.1 | NP_001186274.1 | 5 | 433 | |||
GenBank mRNA | AK304659.1 | - | 5 | 347 | ||
AK311828.1 | - | 6 | 388 | |||
BC000502.2 | - | 5 | 626 | 2821464 | ||
BC017831.1 | - | 1 | 392 | 4251433 | ||
BC066323.1 | - | 1 | 388 | 4797583 | ||
BC066324.1 | - | 1 | 392 | 4799029 | ||
BC106031.1 | - | 1 | 359 | 4540700 | ||
BG758380.1 | - | 7 | 590 | |||
BG827384.1 | - | 6 | 474 | |||
BI086293.1 | - | 6 | 445 | |||
BQ877039.1 | - | 7 | 612 | |||
BQ882430.1 | - | 6 | 538 | |||
CD386294.1 | - | 6 | 493 | |||
CD387752.1 | - | 6 | 578 | |||
X53777.1 | - | 5 | 491 |
Gene Symbol: RPL17-C18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199355.1 | NP_001186284.1 | 5 | 366 | ||
NM_001199356.1 | NP_001186285.1 | 5 | 433 | |||
GenBank mRNA | BI461805.1 | - | 5 | 352 | ||
BX393840.2 | - | 5 | 368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
