siRNA Details
Gene Symbol: CSHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318.3 | NP_001309.3 | 3 | 381 | ||
NM_001321067.1 | NP_001307996.1 | 3 | 377 | |||
NM_001321068.1 | NP_001307997.1 | 4 | 371 | |||
NM_001321069.1 | NP_001307998.1 | 4 | 440 | |||
NM_022579.2 | NP_072101.1 | 4 | 557 | |||
NM_022580.2 | NP_072102.1 | 3 | 308 | |||
NM_022581.2 | NP_072103.1 | 4 | 488 | |||
XM_011524344.1 | XP_011522646.1 | 4 | 487 | |||
XM_011524346.2 | XP_011522648.1 | 3 | 329 | |||
GenBank mRNA | AX772828.1 | - | 4 | 385 | ||
BC029365.1 | - | 3 | 311 | 4619624 | ||
BX382469.2 | - | 3 | 381 | |||
DR004701.1 | - | 4 | 281 |
Gene Symbol: CSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317.5 | NP_001308.1 | 4 | 560 | ||
GenBank mRNA | AK291587.1 | - | 4 | 479 | ||
AU134786.2 | - | 4 | 560 | |||
BC002717.2 | - | 4 | 453 | 3631916 | ||
BC005921.1 | - | 4 | 472 | 4043768 | ||
BC020756.1 | - | 4 | 528 | 4769769 | ||
BC057768.1 | - | 4 | 472 | 30337725 | ||
BC062775.1 | - | 4 | 476 | 4604848 | ||
BG436049.1 | - | 4 | 473 | |||
BT006926.1 | - | 3 | 410 | |||
J00118.1 | - | 4 | 439 | |||
V00593.1 | - | 3 | 262 |
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020991.3 | NP_066271.1 | 4 | 525 | ||
NM_022644.3 | NP_072170.1 | 4 | 525 | |||
GenBank mRNA | BC022044.1 | - | 4 | 476 | 4769046 | |
BC035965.1 | - | 4 | 472 | 4731880 | ||
BC119748.2 | - | 4 | 469 | 40119242 | ||
V00573.1 | - | 4 | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
