siRNA Details
Gene Symbol: TIAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004740.3 | NP_004731.2 | 1 | 238 | ||
GenBank mRNA | AK054958.1 | - | 1 | 238 | ||
AK126861.1 | - | 1 | 1935 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
