siRNA Details
Gene Symbol: TIAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004740.3 | NP_004731.2 | 1 | 1637 | ||
GenBank mRNA | AF105277.1 | - | 1 | 346 | ||
AK054958.1 | - | 1 | 1637 | |||
AK126861.1 | - | 1 | 3334 | |||
BC047669.1 | - | 1 | 934 | 6056136 | ||
BC058075.1 | - | 1 | 620 | |||
BC131561.1 | - | 1 | 834 | 40109188 |
Gene Symbol: MYO18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078471.3 | NP_510880.2 | 42 | 7120 | ||
NM_203318.1 | NP_976063.1 | 41 | 7075 | |||
GenBank mRNA | AB177858.1 | - | 41 | 6939 | ||
AB177860.1 | - | 41 | 6027 | |||
AF252258.1 | - | 4 | 1373 | |||
AK122870.1 | - | 8 | 2211 | |||
AY703984.1 | - | 41 | 6027 | |||
BC039612.1 | - | 40 | 7068 | 6045482 | ||
CR933614.1 | - | 41 | 7049 | |||
D86970.2 | - | 42 | 7143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
