siRNA Details
Gene Symbol: HSD17B13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136230.2 | NP_001129702.1 | 6 | 1511 | ||
NM_178135.4 | NP_835236.2 | 7 | 1619 | |||
GenBank mRNA | AB073347.1 | - | 7 | 1588 | ||
AY186249.1 | - | 7 | 1599 | |||
AY186250.1 | - | 6 | 1491 | |||
AY358575.1 | - | 7 | 1619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
