siRNA Details
Gene Symbol: RGPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005054.2 | NP_005045.2 | 23 | 7038 | ||
XM_006712794.3 | XP_006712857.2 | 23 | 7069 | |||
XM_017005080.1 | XP_016860569.1 | 23 | 7058 | |||
XM_017005082.1 | XP_016860571.1 | 21 | 6740 | |||
GenBank mRNA | AB209082.1 | - | 4 | 3937 | ||
CR936614.1 | - | 3 | 6763 | |||
EU794682.1 | - | 23 | 7038 |
Gene Symbol: GCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181453.3 | NP_852118.1 | 23 | 6850 | ||
NR_028063.1 | 22 | 6765 | ||||
XM_006712870.3 | XP_006712933.1 | 23 | 7419 | |||
XM_006712871.1 | XP_006712934.1 | 22 | 6673 | |||
XM_011512213.2 | XP_011510515.1 | 22 | 6666 | |||
GenBank mRNA | AB002334.1 | - | 22 | 6647 | ||
BC146789.1 | - | 22 | 6647 | 9007253 |
Gene Symbol: RGPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123363.3 | NP_001116835.1 | 24 | 7494 | ||
XM_011511766.2 | XP_011510068.1 | 23 | 7078 | |||
XM_017004842.1 | XP_016860331.1 | 23 | 7058 | |||
XM_017004844.1 | XP_016860333.1 | 21 | 6740 |
Gene Symbol: RGPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164463.1 | NP_001157935.1 | 23 | 7153 | ||
GenBank mRNA | AF012086.1 | - | 4 | 3931 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
