siRNA Details
Gene Symbol: RGPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005054.2 | NP_005045.2 | 19, 20 | 2782 | ||
XM_006712794.3 | XP_006712857.2 | 19, 20 | 2813 | |||
XM_017005080.1 | XP_016860569.1 | 19, 20 | 2802 | |||
XM_017005081.1 | XP_016860570.1 | 19, 20 | 2813 | |||
XM_017005082.1 | XP_016860571.1 | 17, 18 | 2484 | |||
GenBank mRNA | AL044843.1 | - | 1, 2 | 89 | ||
EU794682.1 | - | 19, 20 | 2782 |
Gene Symbol: RANBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005264007.2 | XP_005264064.1 | 19, 20 | 2794 | ||
XM_011511578.2 | XP_011509880.1 | 19, 20 | 2836 |
Gene Symbol: RGPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024457.3 | NP_001019628.3 | 19, 20 | 2676 | ||
XM_011532845.2 | XP_011531147.1 | 20, 21 | 2875 | |||
XM_011532849.2 | XP_011531151.1 | 20, 21 | 2875 | |||
XM_011532850.1 | XP_011531152.1 | 20, 21 | 2875 | |||
XM_011532851.2 | XP_011531153.1 | 20, 21 | 2875 | |||
XM_011532852.2 | XP_011531154.1 | 20, 21 | 2875 | |||
XM_017004103.1 | XP_016859592.1 | 20, 21 | 2875 | |||
XM_017004104.1 | XP_016859593.1 | 19, 20 | 2992 | |||
XM_017004105.1 | XP_016859594.1 | 19, 20 | 2840 | |||
XM_017004106.1 | XP_016859595.1 | 19, 20 | 2959 | |||
XM_017004107.1 | XP_016859596.1 | 20, 21 | 2875 | |||
GenBank mRNA | AK303443.1 | - | 5, 6 | 592 | ||
CR749330.1 | - | 19, 20 | 2676 |
Gene Symbol: RGPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078170.2 | NP_001071638.2 | 19, 20 | 2722 | ||
XM_017004845.1 | XP_016860334.1 | 23, 24 | 3173 | |||
XM_017004846.1 | XP_016860335.1 | 19, 20 | 2844 | |||
XM_017004847.1 | XP_016860336.1 | 19, 20 | 2841 | |||
XM_017004848.1 | XP_016860337.1 | 23, 24 | 3173 | |||
XM_017004849.1 | XP_016860338.1 | 23, 24 | 3173 | |||
XM_017004850.1 | XP_016860339.1 | 23, 24 | 3173 | |||
GenBank mRNA | AK300352.1 | - | 8, 9 | 1062 | ||
AY354203.1 | - | 6, 7 | 698 |
Gene Symbol: RGPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144013.1 | NP_001137485.1 | 19, 20 | 2765 | ||
XM_017004738.1 | XP_016860227.1 | 20, 21 | 2814 | |||
XM_017004739.1 | XP_016860228.1 | 20, 21 | 2814 | |||
GenBank mRNA | AK310529.1 | - | 5, 6 | 544 |
Gene Symbol: RGPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182588.2 | NP_872394.2 | 19, 20 | 2790 | ||
XM_011511017.2 | XP_011509319.1 | 19, 20 | 2737 | |||
XM_011511018.2 | XP_011509320.1 | 19, 20 | 2740 | |||
XM_011511019.2 | XP_011509321.1 | 19, 20 | 2736 | |||
XM_011511020.2 | XP_011509322.1 | 19, 20 | 2736 | |||
GenBank mRNA | AK302921.1 | - | 15, 16 | 2098 | ||
BX537861.1 | - | 14, 15 | 3688 |
Gene Symbol: RGPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123363.3 | NP_001116835.1 | 20, 21 | 3238 | ||
XM_011511766.2 | XP_011510068.1 | 19, 20 | 2822 | |||
XM_011511767.2 | XP_011510069.1 | 19, 20 | 2822 | |||
XM_011511769.2 | XP_011510071.1 | 19, 20 | 2822 | |||
XM_011511770.2 | XP_011510072.1 | 19, 20 | 2822 | |||
XM_017004839.1 | XP_016860328.1 | 19, 20 | 2822 | |||
XM_017004840.1 | XP_016860329.1 | 19, 20 | 2895 | |||
XM_017004841.1 | XP_016860330.1 | 19, 20 | 3025 | |||
XM_017004842.1 | XP_016860331.1 | 19, 20 | 2802 | |||
XM_017004843.1 | XP_016860332.1 | 19, 20 | 2822 | |||
XM_017004844.1 | XP_016860333.1 | 17, 18 | 2484 | |||
GenBank mRNA | AK056675.1 | - | 15, 16 | 2096 |
Gene Symbol: RGPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164463.1 | NP_001157935.1 | 19, 20 | 2897 | ||
XM_005263747.4 | XP_005263804.1 | 19, 20 | 2895 | |||
XM_011511733.2 | XP_011510035.1 | 19, 20 | 2895 | |||
XM_017004825.1 | XP_016860314.1 | 19, 20 | 2895 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
