siRNA Details
Gene Symbol: HSFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033108.2 | NP_149099.2 | 2 | 1394 | ||
XM_005262565.3 | XP_005262622.1 | 3 | 1265 | |||
XM_017030085.1 | XP_016885574.1 | 3 | 1208 | |||
GenBank mRNA | AF332227.1 | - | 2 | 1391 | ||
BC036567.2 | - | 2 | 1394 | 5297256 |
Gene Symbol: HSFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153716.1 | NP_714927.1 | 2 | 1411 | ||
XM_005262508.3 | XP_005262565.1 | 3 | 1313 | |||
XM_017030030.1 | XP_016885519.1 | 3 | 1213 | |||
GenBank mRNA | AK058182.1 | - | 2 | 1411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
