siRNA Details
Gene Symbol: LCE1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178352.2 | NP_848129.1 | 2 | 389 | ||
GenBank mRNA | BG697954.1 | - | 2 | 389 |
Gene Symbol: LCE1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178353.1 | NP_848130.1 | 2 | 411 | ||
GenBank mRNA | BC038391.1 | - | 2 | 411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
