siRNA Details
Gene Symbol: UGT1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001072.3 | NP_001063.2 | 5 | 1765 | ||
NM_205862.1 | NP_995584.1 | 6 | 978 | |||
GenBank mRNA | AY435141.1 | - | 5 | 1626 | ||
BC019861.1 | - | 5 | 1660 | 4454322 |
Gene Symbol: UGT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000463.2 | NP_000454.1 | 5 | 1644 | ||
GenBank mRNA | AK290834.1 | - | 5 | 1648 | ||
AY435136.1 | - | 5 | 1629 | |||
BC128414.1 | - | 5 | 1645 | 40117598 | ||
BC128415.1 | - | 5 | 1644 | 40117599 | ||
JQ686667.1 | - | 5 | 1629 | |||
JQ699637.1 | - | 5 | 1629 | |||
JQ699638.1 | - | 5 | 1629 | |||
JQ699639.1 | - | 5 | 1629 | |||
JQ699640.1 | - | 5 | 1629 | |||
JQ699641.1 | - | 5 | 1629 | |||
JQ699642.1 | - | 5 | 1629 | |||
JQ699643.1 | - | 5 | 1629 | |||
JQ699644.1 | - | 5 | 1629 | |||
JQ699645.1 | - | 5 | 1629 | |||
JQ699646.1 | - | 5 | 1629 | |||
JQ699647.1 | - | 5 | 1629 | |||
JQ699649.1 | - | 5 | 1629 | |||
JQ699650.1 | - | 5 | 1629 | |||
M57899.1 | - | 5 | 1644 |
Gene Symbol: UGT1A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019075.2 | NP_061948.1 | 5 | 1666 | ||
GenBank mRNA | AK025403.1 | - | 5 | 1273 | ||
AY435137.1 | - | 5 | 1620 | |||
BC020971.1 | - | 5 | 1689 | 3850654 | ||
BC069210.1 | - | 5 | 1650 | 4660540 |
Gene Symbol: UGT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019093.2 | NP_061966.1 | 5 | 1632 | ||
GenBank mRNA | AY435138.1 | - | 5 | 1632 |
Gene Symbol: UGT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007120.2 | NP_009051.1 | 5 | 1661 | ||
GenBank mRNA | AY435139.1 | - | 5 | 1632 | ||
BC121036.1 | - | 7 | 1766 | 40117585 | ||
BC131623.1 | - | 5 | 1625 | |||
BC139784.1 | - | 5 | 1652 | 40117089 | ||
M57951.1 | - | 5 | 1661 |
Gene Symbol: UGT1A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019078.1 | NP_061951.1 | 5 | 1632 | ||
GenBank mRNA | AY435140.1 | - | 5 | 1632 |
Gene Symbol: UGT1A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019077.2 | NP_061950.2 | 5 | 1620 | ||
GenBank mRNA | AY435142.1 | - | 5 | 1620 |
Gene Symbol: UGT1A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019076.4 | NP_061949.3 | 5 | 1683 | ||
GenBank mRNA | AF462267.1 | - | 5 | 1653 | ||
AF462268.1 | - | 5 | 1683 | |||
AY435143.1 | - | 5 | 1620 |
Gene Symbol: UGT1A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021027.2 | NP_066307.1 | 5 | 1657 | ||
GenBank mRNA | AF056188.1 | - | 5 | 1638 | ||
AY435144.1 | - | 5 | 1620 | |||
BC058844.1 | - | 5 | 1663 | 4593613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
