siRNA Details
Gene Symbol: UGT1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001072.3 | NP_001063.2 | 2 | 1066 | ||
NM_205862.1 | NP_995584.1 | 3 | 279 | |||
GenBank mRNA | AY435141.1 | - | 2 | 927 | ||
BC019861.1 | - | 2 | 961 | 4454322 | ||
BM688740.1 | - | 3 | 279 | |||
BM924331.1 | - | 3 | 297 | |||
DQ364250.1 | - | 2 | 935 | |||
J04093.1 | - | 2 | 1011 |
Gene Symbol: UGT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000463.2 | NP_000454.1 | 2 | 945 | ||
GenBank mRNA | AK290834.1 | - | 2 | 949 | ||
AY435136.1 | - | 2 | 930 | |||
BC128414.1 | - | 2 | 946 | 40117598 | ||
BC128415.1 | - | 2 | 945 | 40117599 | ||
DQ364247.1 | - | 2 | 930 | |||
JQ686667.1 | - | 2 | 930 | |||
JQ699637.1 | - | 2 | 930 | |||
JQ699638.1 | - | 2 | 930 | |||
JQ699639.1 | - | 2 | 930 | |||
JQ699640.1 | - | 2 | 930 | |||
JQ699641.1 | - | 2 | 930 | |||
JQ699642.1 | - | 2 | 930 | |||
JQ699643.1 | - | 2 | 930 | |||
JQ699644.1 | - | 2 | 930 | |||
JQ699645.1 | - | 2 | 930 | |||
JQ699646.1 | - | 2 | 930 | |||
JQ699647.1 | - | 2 | 930 | |||
JQ699649.1 | - | 2 | 930 | |||
JQ699650.1 | - | 2 | 930 | |||
M57899.1 | - | 2 | 945 |
Gene Symbol: UGT1A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019075.2 | NP_061948.1 | 2 | 967 | ||
GenBank mRNA | AK025403.1 | - | 2 | 574 | ||
AY435137.1 | - | 2 | 921 | |||
BC020971.1 | - | 2 | 990 | 3850654 | ||
BC053576.1 | - | 2 | 942 | 4452440 | ||
BC069210.1 | - | 2 | 951 | 4660540 | ||
U89508.1 | - | 2 | 921 |
Gene Symbol: UGT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019093.2 | NP_061966.1 | 2 | 933 | ||
GenBank mRNA | AY435138.1 | - | 2 | 933 | ||
DQ364248.1 | - | 2 | 201 |
Gene Symbol: UGT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007120.2 | NP_009051.1 | 2 | 962 | ||
GenBank mRNA | AK313623.1 | - | 2 | 976 | ||
AY435139.1 | - | 2 | 933 | |||
BC121036.1 | - | 3 | 933 | 40117585 | ||
BC131623.1 | - | 2 | 926 | |||
BC139784.1 | - | 2 | 953 | 40117089 | ||
DQ364249.1 | - | 2 | 942 | |||
M57951.1 | - | 2 | 962 |
Gene Symbol: UGT1A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019078.1 | NP_061951.1 | 2 | 933 | ||
GenBank mRNA | AY435140.1 | - | 2 | 933 | ||
DQ383514.1 | - | 2 | 642 |
Gene Symbol: UGT1A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019077.2 | NP_061950.2 | 2 | 921 | ||
GenBank mRNA | AY435142.1 | - | 2 | 921 | ||
DQ383513.1 | - | 2 | 633 | |||
U89507.1 | - | 2 | 921 |
Gene Symbol: UGT1A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019076.4 | NP_061949.3 | 2 | 984 | ||
GenBank mRNA | AF030310.1 | - | 2 | 921 | ||
AF462267.1 | - | 2 | 954 | |||
AF462268.1 | - | 2 | 984 | |||
AK313488.1 | - | 2 | 967 | |||
AY435143.1 | - | 2 | 921 | |||
DQ364251.1 | - | 2 | 576 |
Gene Symbol: UGT1A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021027.2 | NP_066307.1 | 2 | 958 | ||
GenBank mRNA | AF056188.1 | - | 2 | 939 | ||
AK313510.1 | - | 2 | 997 | |||
AY435144.1 | - | 2 | 921 | |||
BC058844.1 | - | 2 | 964 | 4593613 | ||
DQ364246.1 | - | 2 | 273 | |||
S55985.1 | - | 2 | 955 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
