siRNA Details
Gene Symbol: AXIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004655.3 | NP_004646.3 | 11 | 2916 | ||
XM_011525319.2 | XP_011523621.1 | 11 | 3127 | |||
XM_011525320.1 | XP_011523622.1 | 11 | 3116 | |||
XM_011525321.2 | XP_011523623.1 | 11 | 3064 | |||
XM_017025192.1 | XP_016880681.1 | 11 | 2783 | |||
XM_017025193.1 | XP_016880682.1 | 10 | 2921 | |||
XM_017025194.1 | XP_016880683.1 | 10 | 2700 | |||
GenBank mRNA | AB052751.1 | - | 4 | 541 | ||
AF078165.1 | - | 10 | 2716 | |||
AK025718.1 | - | 10 | 1938 | |||
CR933657.1 | - | 11 | 2941 | |||
CX759580.1 | - | 2 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
