siRNA Details
Gene Symbol: AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004065.4 | NP_001004065.2 | 4 | 5711 | ||
NM_001136562.2 | NP_001130034.1 | 4 | 5549 | |||
NM_001198656.1 | NP_001185585.1 | 5 | 5750 | |||
GenBank mRNA | AJ420463.1 | - | 1 | 61 | ||
BC010513.1 | - | 1 | 345 | |||
CN302358.1 | - | 1 | 183 |
Gene Symbol: PALM2-AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007203.4 | NP_009134.1 | 11 | 6266 | ||
NM_147150.2 | NP_671492.1 | 10 | 6227 | |||
GenBank mRNA | AJ303079.1 | - | 11 | 6257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
