siRNA Details
Gene Symbol: SLC44A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130058.1 | NP_001123530.1 | 24 | 2489 | ||
NM_001320285.1 | NP_001307214.1 | 26 | 2649 | |||
NM_001320287.1 | NP_001307216.1 | 25 | 2582 | |||
XM_006710445.3 | XP_006710508.1 | 22 | 5262 | |||
XM_011540986.2 | XP_011539288.1 | 23 | 5353 | |||
XM_011540987.2 | XP_011539289.1 | 25 | 5430 | |||
XM_017000607.1 | XP_016856096.1 | 23 | 5401 | |||
XM_017000608.1 | XP_016856097.1 | 23 | 2514 | |||
XM_017000609.1 | XP_016856098.1 | 26 | 6719 | |||
XM_017000610.1 | XP_016856099.1 | 26 | 5797 | |||
XM_017000611.1 | XP_016856100.1 | 24 | 5482 | |||
GenBank mRNA | AK128063.1 | - | 25 | 2542 | ||
BC034580.1 | - | 26 | 2628 | 4824861 | ||
BC051740.1 | - | 26 | 2628 | 4824908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
