siRNA Details
Gene Symbol: SPANXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145662.3 | NP_663695.1 | 2 | 384 | ||
GenBank mRNA | CB110020.1 | - | 2 | 384 | ||
CB113891.1 | - | 2 | 384 |
Gene Symbol: SPANXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013453.3 | NP_038481.2 | 2 | 384 | ||
GenBank mRNA | AF098306.1 | - | 2 | 360 | ||
AJ238277.1 | - | 2 | 363 | |||
AK225988.1 | - | 2 | 383 | |||
BC054023.1 | - | 2 | 385 | 6648369 | ||
BC069393.1 | - | 2 | 360 | 7262682 | ||
BC069816.1 | - | 2 | 360 | 7262670 | ||
CB115288.1 | - | 2 | 384 |
Gene Symbol: SPANXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022661.3 | NP_073152.2 | 2 | 417 |
Gene Symbol: SPANXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ457795.1 | - | 2 | 360 | ||
BC005382.1 | - | 2 | 417 | 3935644 | ||
BC062672.1 | - | 2 | 598 | 4047937 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
