siRNA Details
Gene Symbol: SPANXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013453.3 | NP_038481.2 | 2 | 214 | ||
GenBank mRNA | AF098306.1 | - | 2 | 190 | ||
AJ238277.1 | - | 2 | 193 | |||
AK225988.1 | - | 2 | 213 | |||
AY920732.1 | - | 2 | 154 | |||
AY920733.1 | - | 2 | 154 | |||
AY920735.1 | - | 2 | 154 | |||
AY920736.1 | - | 2 | 154 | |||
AY920737.1 | - | 2 | 154 | |||
AY920738.1 | - | 2 | 154 | |||
AY920739.1 | - | 2 | 154 | |||
AY920741.1 | - | 2 | 154 | |||
AY920742.1 | - | 2 | 154 | |||
AY920743.1 | - | 2 | 154 | |||
AY920744.1 | - | 2 | 154 | |||
AY920745.1 | - | 2 | 154 | |||
AY920746.1 | - | 2 | 154 | |||
BC054023.1 | - | 2 | 215 | 6648369 | ||
BC069393.1 | - | 2 | 190 | 7262682 | ||
BC069816.1 | - | 2 | 190 | 7262670 | ||
CB115288.1 | - | 2 | 214 |
Gene Symbol: SPANXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145662.3 | NP_663695.1 | 2 | 214 | ||
GenBank mRNA | CB110020.1 | - | 2 | 214 | ||
CB113891.1 | - | 2 | 214 |
Gene Symbol: SPANXB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032461.3 | NP_115850.2 | 2 | 275 | ||
GenBank mRNA | AF098307.1 | - | 2 | 275 | ||
AJ457796.1 | - | 2 | 275 | |||
BC034472.1 | - | 2 | 273 | 4824918 | ||
BC137533.1 | - | 2 | 265 | 9021533 | ||
BC137536.1 | - | 2 | 265 | 9021536 |
Gene Symbol: SPANXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022661.3 | NP_073152.2 | 2 | 247 | ||
GenBank mRNA | AA412605.1 | - | 2 | 210 |
Gene Symbol: SPANXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032417.3 | NP_115793.1 | 2 | 488 | ||
GenBank mRNA | AF312765.1 | - | 2 | 164 | ||
AJ457795.1 | - | 2 | 190 | |||
AY920734.1 | - | 2 | 154 | |||
AY920740.1 | - | 2 | 154 | |||
BC005382.1 | - | 2 | 247 | 3935644 | ||
BC062672.1 | - | 2 | 428 | 4047937 | ||
BC103961.1 | - | 2 | 187 | 40013988 | ||
BC121070.2 | - | 2 | 197 | 40119569 | ||
BC134418.1 | - | 2 | 197 | 40119568 | ||
BM554791.1 | - | 2 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
