siRNA Details
Gene Symbol: POMZP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012230.3 | NP_036362.3 | 2 | 613 | ||
NM_152992.2 | NP_694537.1 | 2 | 613 | |||
GenBank mRNA | U10099.1 | - | 2 | 559 |
Gene Symbol: POM121C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099415.2 | NP_001092885.2 | 4 | 748 | ||
GenBank mRNA | AB354586.1 | - | 2 | 592 | ||
BC082993.1 | - | 4 | 731 | |||
BC130587.1 | - | 4 | 455 | |||
BC130589.1 | - | 4 | 455 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
