siRNA Details
Gene Symbol: LRRC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024678.3 | NP_001019849.2 | 4 | 588 | ||
GenBank mRNA | AB178281.1 | - | 3 | 519 | ||
BC111067.1 | - | 4 | 580 | 5767723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
