siRNA Details
Gene Symbol: C22orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182520.2 | NP_872326.2 | 3, 4 | 536 | ||
XM_011529907.2 | XP_011528209.1 | 2, 3 | 1327 | |||
XM_017028612.1 | XP_016884101.1 | 3, 4 | 1032 | |||
XM_017028614.1 | XP_016884103.1 | 2, 3 | 1327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
