siRNA Details
Gene Symbol: POLR2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032959.5 | NP_116581.3 | 5 | 651 | ||
GenBank mRNA | BC056864.1 | - | 5 | 541 | ||
BC086857.1 | - | 4 | 476 | 6007730 |
Gene Symbol: LOC107983993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012895.1 | XP_016868384.1 | 3 | 370 | ||
XM_017012896.1 | XP_016868385.1 | 3 | 893 |
Gene Symbol: POLR2J3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097615.2 | NP_001091084.2 | 5 | 623 | ||
XM_017012373.1 | XP_016867862.1 | 6 | 817 | |||
GenBank mRNA | BC113031.1 | - | 2 | 680 | ||
DA574442.1 | - | 4 | 411 |
Gene Symbol: POLR2J4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003655.3 | 5 | 610 | |||
GenBank mRNA | AK296066.1 | - | 4 | 413 | ||
BX537878.1 | - | 5 | 565 | |||
CD674494.1 | - | 3 | 307 |
Gene Symbol: UPK3BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114403.2 | NP_001107875.1 | 2 | 283 | ||
GenBank mRNA | EU341824.1 | - | 2 | 283 | ||
KF150200.1 | - | 2 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
