siRNA Details
Gene Symbol: WDFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014991.4 | NP_055806.2 | 68 | 12419 | ||
XM_005262858.4 | XP_005262915.1 | 70 | 12460 | |||
XM_011531757.2 | XP_011530059.1 | 69 | 12371 | |||
XM_011531759.2 | XP_011530061.1 | 68 | 12333 | |||
XM_011531760.2 | XP_011530062.1 | 68 | 12271 | |||
XM_011531761.2 | XP_011530063.1 | 69 | 12326 | |||
XM_011531762.2 | XP_011530064.1 | 68 | 12320 | |||
XM_011531763.2 | XP_011530065.1 | 67 | 12217 | |||
XM_011531764.2 | XP_011530066.1 | 68 | 12287 | |||
XM_011531765.2 | XP_011530067.1 | 65 | 12022 | |||
XM_017007906.1 | XP_016863395.1 | 71 | 12588 | |||
XM_017007907.1 | XP_016863396.1 | 71 | 12585 | |||
XM_017007908.1 | XP_016863397.1 | 70 | 12517 | |||
XM_017007909.1 | XP_016863398.1 | 56 | 10213 | |||
GenBank mRNA | AB023210.2 | - | 33 | 6102 | ||
BC150246.1 | - | 33 | 6123 | |||
BX648077.1 | - | 25 | 4984 | |||
BX648153.1 | - | 28 | 5314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
