siRNA Details
Gene Symbol: WDFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014991.4 | NP_055806.2 | 65 | 10259 | ||
XM_005262858.4 | XP_005262915.1 | 67 | 10300 | |||
XM_011531757.2 | XP_011530059.1 | 66 | 10211 | |||
XM_011531759.2 | XP_011530061.1 | 65 | 10173 | |||
XM_011531760.2 | XP_011530062.1 | 65 | 10111 | |||
XM_011531761.2 | XP_011530063.1 | 66 | 10166 | |||
XM_011531762.2 | XP_011530064.1 | 65 | 10160 | |||
XM_011531763.2 | XP_011530065.1 | 64 | 10057 | |||
XM_011531764.2 | XP_011530066.1 | 65 | 10127 | |||
XM_011531765.2 | XP_011530067.1 | 62 | 9862 | |||
XM_017007906.1 | XP_016863395.1 | 68 | 10428 | |||
XM_017007907.1 | XP_016863396.1 | 68 | 10425 | |||
XM_017007908.1 | XP_016863397.1 | 67 | 10357 | |||
XM_017007909.1 | XP_016863398.1 | 53 | 8053 | |||
GenBank mRNA | AB023210.2 | - | 30 | 3942 | ||
AF538685.1 | - | 64 | 10151 | |||
AK092394.1 | - | 5 | 638 | |||
BC013377.2 | - | 2 | 603 | |||
BC015214.1 | - | 5 | 585 | |||
BC065502.1 | - | 12 | 1609 | |||
BC119633.2 | - | 2 | 306 | |||
BC148294.1 | - | 2 | 306 | |||
BC148298.2 | - | 2 | 306 | |||
BC150246.1 | - | 30 | 3963 | |||
BX648077.1 | - | 22 | 2824 | |||
BX648153.1 | - | 25 | 3152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
