siRNA Details
Gene Symbol: WDFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014991.4 | NP_055806.2 | 14 | 2583 | ||
XM_005262858.4 | XP_005262915.1 | 16 | 2624 | |||
XM_011531757.2 | XP_011530059.1 | 15 | 2535 | |||
XM_011531759.2 | XP_011530061.1 | 14 | 2497 | |||
XM_011531760.2 | XP_011530062.1 | 14 | 2435 | |||
XM_011531761.2 | XP_011530063.1 | 15 | 2535 | |||
XM_011531762.2 | XP_011530064.1 | 15 | 2535 | |||
XM_011531763.2 | XP_011530065.1 | 13 | 2381 | |||
XM_011531764.2 | XP_011530066.1 | 15 | 2535 | |||
XM_011531765.2 | XP_011530067.1 | 12 | 2237 | |||
XM_011531766.2 | XP_011530068.1 | 15 | 2535 | |||
XM_011531767.2 | XP_011530069.1 | 15 | 2535 | |||
XM_017007906.1 | XP_016863395.1 | 17 | 2752 | |||
XM_017007907.1 | XP_016863396.1 | 17 | 2749 | |||
XM_017007908.1 | XP_016863397.1 | 16 | 2681 | |||
XM_017007909.1 | XP_016863398.1 | 2 | 377 | |||
GenBank mRNA | AF538685.1 | - | 13 | 2475 | ||
AK092019.1 | - | 12 | 2237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
