siRNA Details
Gene Symbol: KLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030047.1 | NP_001025218.1 | 3 | 525 | ||
NM_001030048.1 | NP_001025219.1 | 3 | 396 | |||
NM_001648.2 | NP_001639.1 | 3 | 525 | |||
GenBank mRNA | AF335477.1 | - | 3 | 948 | ||
AF335478.1 | - | 2 | 1763 | |||
AJ459782.1 | - | 3, 4 | 382 | |||
AJ459783.1 | - | 3 | 505 | |||
AJ512346.1 | - | 3 | 505 | |||
AK312933.1 | - | 3 | 519 | |||
BC005307.1 | - | 3 | 525 | 3950475 | ||
BC050595.2 | - | 3 | 548 | 6199928 | ||
BC056665.1 | - | 3 | 530 | 4296876 | ||
BQ932072.1 | - | 3 | 362 | |||
BT019862.1 | - | 3 | 484 | |||
DQ641251.1 | - | 1 | 916 | |||
M21895.1 | - | 3 | 473 | |||
M21896.1 | - | 3 | 473 | |||
M26663.1 | - | 3 | 508 | |||
S75755.1 | - | 3 | 525 | |||
U17040.1 | - | 3 | 525 | |||
X05332.1 | - | 3 | 527 | |||
X07730.1 | - | 2 | 789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
