siRNA Details
Gene Symbol: CYHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138496.1 | NP_612505.1 | 5 | 1316 | ||
XM_006716562.3 | XP_006716625.2 | 4 | 1542 | |||
XM_011517051.1 | XP_011515353.1 | 4 | 4425 | |||
XM_017013463.1 | XP_016868952.1 | 5 | 1563 | |||
XM_017013464.1 | XP_016868953.1 | 5 | 1332 | |||
XM_017013465.1 | XP_016868954.1 | 5 | 1450 | |||
XM_017013466.1 | XP_016868955.1 | 5 | 1332 | |||
GenBank mRNA | AK094514.1 | - | 5 | 1316 | ||
AK160381.1 | - | 3 | 5119 | |||
BC004544.2 | - | 3 | 932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
