siRNA Details
Gene Symbol: CYHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138496.1 | NP_612505.1 | 3, 4 | 627 | ||
XM_006716562.3 | XP_006716625.2 | 2, 3 | 853 | |||
XM_011517051.1 | XP_011515353.1 | 2, 3 | 3736 | |||
XM_017013463.1 | XP_016868952.1 | 3, 4 | 874 | |||
XM_017013464.1 | XP_016868953.1 | 3, 4 | 643 | |||
XM_017013465.1 | XP_016868954.1 | 3, 4 | 761 | |||
XM_017013466.1 | XP_016868955.1 | 3, 4 | 643 | |||
GenBank mRNA | AB007965.1 | - | 1, 2 | 4716 | ||
AK094514.1 | - | 3, 4 | 627 | |||
AK160381.1 | - | 1, 2 | 4430 | |||
BC004544.2 | - | 1, 2 | 243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
