siRNA Details
Gene Symbol: AHNAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001620.2 | NP_001611.1 | 5 | 18153 | ||
XM_005274240.1 | XP_005274297.1 | 5 | 18050 | |||
XM_005274241.2 | XP_005274298.1 | 5 | 17971 | |||
XM_005274245.1 | XP_005274302.1 | 6 | 16739 | |||
XM_017018270.1 | XP_016873759.1 | 6 | 17894 | |||
XM_017018271.1 | XP_016873760.1 | 6 | 13196 | |||
XM_017018272.1 | XP_016873761.1 | 6 | 12644 | |||
XM_017018273.1 | XP_016873762.1 | 6 | 12644 | |||
GenBank mRNA | BC017483.1 | - | 1 | 1286 | ||
M80899.1 | - | 1 | 4016 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
