siRNA Details
Gene Symbol: AHNAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024060.3 | NP_076965.2 | 4, 5 | 634 | ||
GenBank mRNA | BC000926.1 | - | 4, 5 | 576 | ||
BC128460.1 | - | 4, 5 | 623 | |||
CN482151.1 | - | 1, 2 | 14 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
