siRNA Details
Gene Symbol: AMY1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008219.1 | NP_001008220.1 | 10, 11 | 1648 | ||
XM_011541263.1 | XP_011539565.1 | 10, 11 | 1569 | |||
XM_017001058.1 | XP_016856547.1 | 10, 11 | 1486 | |||
GenBank mRNA | CB985730.1 | - | 3, 4 | 330 |
Gene Symbol: AMY1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008221.1 | NP_001008222.1 | 10, 11 | 1568 | ||
NM_004038.3 | NP_004029.2 | 10, 11 | 1649 | |||
GenBank mRNA | AK292341.1 | - | 10, 11 | 1567 | ||
AK313275.1 | - | 10, 11 | 1569 | |||
BC063129.1 | - | 10, 11 | 1578 | 30327425 | ||
BC069347.1 | - | 9, 10 | 1354 | 7262297 | ||
BC069463.1 | - | 9, 10 | 1354 | 7262309 | ||
BC070302.1 | - | 10, 11 | 1574 | |||
BC092444.1 | - | 10, 11 | 1545 | 30349929 | ||
BC132985.1 | - | 10, 11 | 1524 | 40147007 | ||
BC132987.1 | - | 10, 11 | 1524 | 40147009 | ||
BC132995.1 | - | 10, 11 | 1523 | 40147017 | ||
BC132997.1 | - | 10, 11 | 1524 | 40147019 | ||
BC144452.1 | - | 10, 11 | 1524 | 9052978 |
Gene Symbol: AMY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008218.1 | NP_001008219.1 | 10, 11 | 1626 | ||
XM_011541262.1 | XP_011539564.1 | 10, 11 | 1569 |
Gene Symbol: AMY2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020978.4 | NP_066188.1 | 11, 12 | 2026 | ||
GenBank mRNA | AK095605.1 | - | 9, 10 | 2577 | ||
AK127047.1 | - | 10, 11 | 3465 | |||
BC011179.1 | - | 11, 12 | 1718 | 4150646 | ||
BC020861.1 | - | 10, 11 | 1490 | 4551745 | ||
M24895.1 | - | 9, 10 | 1400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
