siRNA Details
Gene Symbol: C2CD4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136003.1 | NP_001129475.1 | 1 | 93 | ||
GenBank mRNA | AK293673.1 | - | 1 | 93 | ||
DC305608.1 | - | 1 | 93 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
