siRNA Details
Gene Symbol: C2CD4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136003.1 | NP_001129475.1 | 2 | 1637 | ||
XM_011509055.2 | XP_011507357.1 | 2 | 2100 | |||
XM_016999989.1 | XP_016855478.1 | 2 | 2040 | |||
GenBank mRNA | AK293673.1 | - | 2 | 1637 | ||
BC157847.1 | - | 1 | 1115 | 9057066 | ||
BC171843.1 | - | 1 | 1115 | 9054497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
