siRNA Details
Gene Symbol: ECE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037324.2 | NP_001032401.1 | 8, 9 | 1209 | ||
NM_001100120.1 | NP_001093590.1 | 9, 10 | 1434 | |||
NM_001100121.1 | NP_001093591.1 | 9, 10 | 1296 | |||
NM_014693.3 | NP_055508.3 | 9, 10 | 1545 | |||
GenBank mRNA | AB011176.1 | - | 9, 10 | 1291 | ||
AF192531.1 | - | 9, 10 | 1201 | |||
AF428263.1 | - | 9, 10 | 1473 | |||
AF428264.1 | - | 8, 9 | 1114 | |||
AF521189.1 | - | 9, 10 | 1410 | |||
AK293891.1 | - | 8, 9 | 1139 | |||
AK295270.1 | - | 2, 3 | 346 | |||
AK296541.1 | - | 8, 9 | 1192 | |||
AK309236.1 | - | 6, 7 | 923 | |||
AY359003.1 | - | 8, 9 | 1165 | |||
BC142950.1 | - | 8, 9 | 1209 | 8860334 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177942, NM_177941, NM_139293, NM_177940, AF489571, and others.
Gene Ontology Categories:
