siRNA Details
Gene Symbol: CXorf40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013845.1 | NP_001013867.1 | 1, 2 | 305 | ||
XM_005274698.1 | XP_005274755.1 | 1, 2 | 657 | |||
XM_005274700.1 | XP_005274757.1 | 1, 2 | 657 | |||
XM_011531181.2 | XP_011529483.1 | 1, 2 | 657 | |||
XM_017029586.1 | XP_016885075.1 | 1, 2 | 657 | |||
XM_017029592.1 | XP_016885081.1 | 1, 2 | 657 | |||
XM_017029593.1 | XP_016885082.1 | 1, 2 | 657 | |||
GenBank mRNA | BM847839.1 | - | 1, 2 | 305 | ||
L43575.1 | - | 1, 2 | 231 | |||
U66042.1 | - | 1, 2 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
