siRNA Details
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017940.4 | NP_060410.2 | 3960 | |||
GenBank mRNA | AB051480.1 | - | 17 | 2017 | ||
AF379613.1 | - | 3 | 259 | |||
AF379624.1 | - | 7 | 745 | |||
AF379627.1 | - | 2 | 110 | |||
AF379628.1 | - | 3 | 397 | |||
AY894575.1 | - | 28 | 3842 | |||
BC034418.1 | - | 12 | 1522 | |||
BC094705.1 | - | 10 | 1168 |
Gene Symbol: LOC102724250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546188.2 | XP_011544490.1 | 18 | 11057 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 2176 | |||
NM_001302371.1 | NP_001289300.1 | 2176 | ||||
GenBank mRNA | AF379616.1 | - | 5 | 723 | ||
AF379632.1 | - | 5 | 434 | |||
AF379634.1 | - | 3 | 268 | |||
AF379635.1 | - | 12 | 1439 | |||
AF380580.1 | - | 5 | 430 | |||
AF380582.1 | - | 1 | 68 | |||
AF419616.1 | - | 7 | 647 | |||
AK000726.1 | - | 16 | 2690 | |||
AL050141.1 | - | 4 | 1972 | |||
AY894565.1 | - | 15 | 1988 | |||
BC021111.1 | - | 17 | 2220 | |||
BC063799.1 | - | 17 | 2203 | |||
BX538005.1 | - | 24 | 3423 |
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 18 | 2466 | ||
NM_183372.5 | NP_899228.4 | 20 | 3160 | |||
NR_046188.3 | 21 | 3228 | ||||
NR_110864.1 | 19 | 3152 | ||||
GenBank mRNA | AF419617.1 | - | 19 | 2579 | ||
AK054850.1 | - | 4 | 905 | |||
AK302413.1 | - | 17 | 2302 | |||
AL832622.2 | - | 21 | 3363 | |||
BC136292.1 | - | 20 | 3160 | 9020279 | ||
BX648497.1 | - | 19 | 2951 |
Gene Symbol: NBPF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278141.1 | NP_001265070.1 | 27 | 4171 | ||
GenBank mRNA | AB033071.1 | - | 27 | 3644 | ||
AF131738.1 | - | 2 | 175 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 16 | 2141 | ||
XM_011509395.2 | XP_011507697.1 | 19 | 2641 | |||
GenBank mRNA | AF161426.1 | - | 4 | 318 | ||
AF379626.1 | - | 2 | 84 | |||
AK095459.1 | - | 3 | 167 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 18 | 2490 | ||
NM_173638.4 | NP_775909.2 | 15 | 1960 | |||
XM_017001077.1 | XP_016856566.1 | 13 | 1608 | |||
GenBank mRNA | AK290142.1 | - | 7 | 662 | ||
AK290302.1 | - | 12 | 1419 | |||
AK299360.1 | - | 9 | 1000 | |||
AY894566.1 | - | 12 | 1400 | |||
BC023087.1 | - | 19 | 2321 | 3918932 | ||
DA932476.1 | - | 2 | 94 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 13 | 1601 | ||
GenBank mRNA | AF379633.1 | - | 7 | 733 | ||
AF379636.1 | - | 5 | 533 | |||
AK292666.1 | - | 4 | 354 | |||
AK295639.1 | - | 7 | 1072 |
Gene Symbol: NBPF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278267.1 | NP_001265196.1 | 1496 | |||
GenBank mRNA | AK299612.1 | - | 4 | 310 |
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104217.1 | 19 | 2295 | |||
GenBank mRNA | AK125792.1 | - | 19 | 2295 |
Gene Symbol: NBPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256416.2 | NP_001243345.1 | 15 | 2083 | ||
NM_001256417.2 | NP_001243346.1 | 14 | 1909 | |||
NM_032264.4 | NP_115640.1 | 15 | 2119 | |||
NR_046176.2 | 18 | 2648 | ||||
XM_006710957.1 | XP_006711020.1 | 15 | 1977 | |||
XM_006710958.1 | XP_006711021.1 | 18 | 2433 | |||
XM_006710959.2 | XP_006711022.1 | 16 | 2217 | |||
XM_011542276.1 | XP_011540578.1 | 19 | 2690 | |||
XM_011542277.1 | XP_011540579.1 | 19 | 2675 | |||
XM_011542278.1 | XP_011540580.1 | 19 | 2504 | |||
XM_011542281.1 | XP_011540583.1 | 16 | 2040 | |||
XM_017002498.1 | XP_016857987.1 | 19 | 2489 | |||
XM_017002499.1 | XP_016857988.1 | 14 | 1755 | |||
XM_017002500.1 | XP_016857989.1 | 18 | 2508 | |||
XM_017002501.1 | XP_016857990.1 | 19 | 2393 | |||
XM_017002502.1 | XP_016857991.1 | 18 | 2322 | |||
GenBank mRNA | AK095602.1 | - | 18 | 2602 | ||
AK098438.1 | - | 5 | 364 | |||
AK292580.1 | - | 15 | 1984 | |||
AK299840.1 | - | 14 | 1871 | |||
AK314597.1 | - | 15 | 2047 | |||
AL136890.1 | - | 15 | 1896 | |||
AL834330.1 | - | 11 | 1092 | |||
AY598326.1 | - | 14 | 1721 | |||
AY894564.1 | - | 14 | 1625 | |||
BC024011.2 | - | 15 | 2010 | 4827023 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 16 | 2313 | ||
NR_102404.1 | 15 | 2149 | ||||
NR_102405.1 | 14 | 2046 | ||||
GenBank mRNA | AK300749.1 | - | 8 | 594 | ||
AY894561.1 | - | 9 | 1113 | |||
AY894562.1 | - | 11 | 1338 | |||
AY894563.1 | - | 10 | 1212 | |||
AY894567.1 | - | 14 | 2046 | |||
AY894568.1 | - | 15 | 1944 | |||
AY894569.1 | - | 15 | 1946 | |||
AY894570.1 | - | 11 | 1338 | |||
AY894571.1 | - | 15 | 2210 | |||
AY894572.1 | - | 15 | 2149 | |||
AY894573.1 | - | 15 | 2210 | |||
BC086308.1 | - | 16 | 2156 | 5453529 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277444.1 | NP_001264373.1 | 21 | 2723 | ||
XM_006711317.2 | XP_006711380.1 | 22 | 2951 | |||
GenBank mRNA | AK294414.1 | - | 15 | 1881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
