siRNA Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000515.4 | NP_000506.2 | 2, 3 | 256 | ||
GenBank mRNA | A00469.1 | - | 2, 3 | 219 | ||
AF110644.1 | - | 2, 3 | 190 | |||
AF185611.1 | - | 2, 3 | 198 | |||
AV749353.1 | - | 2, 3 | 256 | |||
AY613431.1 | - | 2, 3 | 242 | |||
AY613432.1 | - | 2, 3 | 242 | |||
BC075012.2 | - | 2, 3 | 207 | 30915404 | ||
BC075013.2 | - | 2, 3 | 207 | 30915617 | ||
BC090045.1 | - | 2, 3 | 223 | 30394115 | ||
V00519.1 | - | 2, 3 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
