siRNA Details
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020991.3 | NP_066271.1 | 5 | 701 | ||
NM_022644.3 | NP_072170.1 | 4 | 954 | |||
NM_022645.2 | NP_072171.1 | 3 | 416 | |||
GenBank mRNA | BC022044.1 | - | 5 | 652 | 4769046 | |
BC035965.1 | - | 5 | 648 | 4731880 | ||
BC119748.2 | - | 5 | 645 | 40119242 | ||
V00573.1 | - | 5 | 610 |
Gene Symbol: CSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317.5 | NP_001308.1 | 5 | 736 | ||
GenBank mRNA | AK291587.1 | - | 5 | 655 | ||
BC002717.2 | - | 5 | 629 | 3631916 | ||
BC005921.1 | - | 5 | 648 | 4043768 | ||
BC020756.1 | - | 5 | 704 | 4769769 | ||
BC057768.1 | - | 5 | 648 | 30337725 | ||
BC062775.1 | - | 5 | 652 | 4604848 | ||
BT006926.1 | - | 4 | 586 | |||
J00118.1 | - | 5 | 615 | |||
M25118.1 | - | 1 | 109 | |||
V00593.1 | - | 4 | 438 |
Gene Symbol: CSHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318.3 | NP_001309.3 | 4 | 557 | ||
NM_001321067.1 | NP_001307996.1 | 4 | 553 | |||
NM_001321068.1 | NP_001307997.1 | 5 | 547 | |||
NM_001321069.1 | NP_001307998.1 | 5 | 616 | |||
NM_022579.2 | NP_072101.1 | 5 | 733 | |||
NM_022580.2 | NP_072102.1 | 4 | 484 | |||
NM_022581.2 | NP_072103.1 | 5 | 664 | |||
XM_011524344.1 | XP_011522646.1 | 4 | 916 | |||
XM_011524346.2 | XP_011522648.1 | 3 | 758 | |||
GenBank mRNA | AX772828.1 | - | 5 | 561 | ||
BC029365.1 | - | 4 | 487 | 4619624 | ||
BC119747.2 | - | 4 | 484 | 40119241 | ||
BX382469.2 | - | 3 | 810 | |||
BX426214.2 | - | 5 | 529 | |||
DR004701.1 | - | 5 | 457 |
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000515.4 | NP_000506.2 | 5 | 662 | ||
NM_022559.3 | NP_072053.1 | 5 | 617 | |||
NM_022560.3 | NP_072054.1 | 4 | 542 | |||
GenBank mRNA | A00469.1 | - | 5 | 625 | ||
AF110644.1 | - | 5 | 458 | |||
AF185611.1 | - | 6 | 490 | |||
AV753220.1 | - | 3 | 482 | |||
AY613431.1 | - | 5 | 648 | |||
AY613432.1 | - | 5 | 648 | |||
BC062475.1 | - | 5 | 582 | 30393581 | ||
BC075012.2 | - | 5 | 613 | 30915404 | ||
BC075013.2 | - | 5 | 613 | 30915617 | ||
BC090045.1 | - | 5 | 629 | 30394115 | ||
CD108592.1 | - | 4 | 508 | |||
V00519.1 | - | 5 | 626 |
Gene Symbol: GH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002059.4 | NP_002050.1 | 5 | 727 | ||
NM_022556.3 | NP_072050.1 | 5 | 682 | |||
NM_022557.3 | NP_072051.1 | 4 | 980 | |||
NM_022558.3 | NP_072052.1 | 5 | 723 | |||
GenBank mRNA | AB451332.1 | - | 4 | 541 | ||
AB451477.1 | - | 4 | 541 | |||
AF006060.1 | - | 5 | 553 | |||
AF006061.1 | - | 4 | 582 | |||
BC020760.1 | - | 5 | 648 | 4720475 | ||
CR541902.1 | - | 4 | 586 | |||
CR541924.1 | - | 4 | 586 | |||
J03756.1 | - | 4 | 900 | |||
M38451.1 | - | 5 | 598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
