siRNA Details
Gene Symbol: CGB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033142.1 | NP_149133.1 | 3 | 692 | ||
XM_017027502.1 | XP_016882991.1 | 6 | 2039 | |||
GenBank mRNA | AK092722.1 | - | 5 | 1762 | ||
AK125108.1 | - | 5 | 3199 |
Gene Symbol: CGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033377.1 | NP_203695.2 | 3 | 544 | ||
XM_017026233.1 | XP_016881722.1 | 2 | 386 | |||
GenBank mRNA | BC107144.1 | - | 3 | 682 | ||
BC126460.1 | - | 3 | 426 |
Gene Symbol: CGB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319065.1 | NP_001305994.1 | 3 | 671 | ||
NM_033378.1 | NP_203696.2 | 3 | 544 | |||
XM_005258479.3 | XP_005258536.1 | 2 | 477 | |||
GenBank mRNA | BC069367.1 | - | 3 | 439 | 7262156 | |
BC107143.1 | - | 3 | 671 |
Gene Symbol: CGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000737.3 | NP_000728.1 | 3 | 731 | ||
GenBank mRNA | AK291552.1 | - | 3 | 689 | ||
BC022796.1 | - | 3 | 669 | 4811294 | ||
BC069526.1 | - | 3 | 336 | 7262120 | ||
BC106059.1 | - | 3 | 677 | 4992804 | ||
BX334905.2 | - | 3 | 736 | |||
J00117.1 | - | 3 | 352 |
Gene Symbol: CGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033043.1 | NP_149032.1 | 3 | 692 | ||
GenBank mRNA | BC006290.2 | - | 3 | 653 | 3960125 | |
BC051378.1 | - | 3 | 653 | 3956670 | ||
BC103970.1 | - | 3 | 423 | 40014677 | ||
BC103971.1 | - | 3 | 419 | 40014679 | ||
BC106723.1 | - | 3 | 337 | 40014469 | ||
BC106724.1 | - | 3 | 336 | 40014470 | ||
BT006890.1 | - | 2 | 327 |
Gene Symbol: CGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033183.2 | NP_149439.1 | 3 | 699 | ||
GenBank mRNA | BC030994.1 | - | 3 | 694 | 4605021 | |
BC103969.1 | - | 3 | 419 | 40014676 | ||
BC128603.1 | - | 3 | 419 | 40002893 | ||
BX380841.2 | - | 3 | 679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
